siRNA Details
| Gene Symbol: FYTTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027840.1 | 1 | 26 | |||
| GenBank mRNA | AK302546.1 | - | 1 | 26 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DC401034.1 | - | 1 | 26 | Chr. 3 - 197750390 - 197756539 [+] on Build GRCh38 | |

FYTTD1
Genomic Map