siRNA Details
Gene Symbol: FYTTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027840.1 | 1, 2 | 436 | |||
GenBank mRNA | AK302546.1 | - | 1, 2 | 436 | ||
AK316308.1 | - | 1, 2 | 66 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FYTTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027840.1 | 1, 2 | 436 | |||
GenBank mRNA | AK302546.1 | - | 1, 2 | 436 | ||
AK316308.1 | - | 1, 2 | 66 |