siRNA Details
| Gene Symbol: FYTTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027840.1 | 1 | 391 | |||
| GenBank mRNA | AK302546.1 | - | 1 | 391 | ||
| AK316308.1 | - | 1 | 21 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FYTTD1| Gene Symbol: FYTTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027840.1 | 1 | 391 | |||
| GenBank mRNA | AK302546.1 | - | 1 | 391 | ||
| AK316308.1 | - | 1 | 21 | |||