siRNA Details
| Gene Symbol: TPTE2P6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002815.2 | 3 | 1740 | |||
| GenBank mRNA | AL122103.1 | - | 1 | 602 | ||
| AL137411.1 | - | 7 | 698 | |||
| BC045761.1 | - | 3 | 1739 | |||
| DN831683.1 | - | 3 | 295 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TPTE2P6| Gene Symbol: TPTE2P6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002815.2 | 3 | 1740 | |||
| GenBank mRNA | AL122103.1 | - | 1 | 602 | ||
| AL137411.1 | - | 7 | 698 | |||
| BC045761.1 | - | 3 | 1739 | |||
| DN831683.1 | - | 3 | 295 | |||