siRNA Details
| Gene Symbol: TPTE2P6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002815.2 | 1 | 1184 | |||
| GenBank mRNA | BC045761.1 | - | 1 | 1183 | ||
| Gene Symbol: TPTE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271850.1 | NP_001258779.1 | 1 | 54 | ||
| GenBank mRNA | FJ969729.1 | - | 1 | 56 | ||

TPTE2P6
Genomic Map