siRNA Details
| Gene Symbol: TPTE2P6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002815.2 | 1 | 1052 | |||
| GenBank mRNA | BC045761.1 | - | 1 | 1051 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TPTE2P6| Gene Symbol: TPTE2P6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002815.2 | 1 | 1052 | |||
| GenBank mRNA | BC045761.1 | - | 1 | 1051 | ||