siRNA Details
| Gene Symbol: TPTE2P6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002815.2 | 7, 8 | 2258 | |||
| GenBank mRNA | AL137411.1 | - | 11, 12 | 1252 | ||
| BC045761.1 | - | 7, 8 | 2257 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TPTE2P6| Gene Symbol: TPTE2P6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002815.2 | 7, 8 | 2258 | |||
| GenBank mRNA | AL137411.1 | - | 11, 12 | 1252 | ||
| BC045761.1 | - | 7, 8 | 2257 | |||