siRNA Details
| Gene Symbol: NXF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002216.1 | 15 | 1669 | |||
| GenBank mRNA | AK124700.1 | - | 15 | 1669 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NXF4| Gene Symbol: NXF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002216.1 | 15 | 1669 | |||
| GenBank mRNA | AK124700.1 | - | 15 | 1669 | ||