siRNA Details
| Gene Symbol: BBIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195304.1 | NP_001182233.1 | 5 | 1772 | ||
| NM_001195305.1 | NP_001182234.1 | 4 | 1615 | |||
| NM_001195306.1 | NP_001182235.1 | 4 | 1461 | |||
| NM_001195307.1 | NP_001182236.1 | 3 | 1540 | |||
| NM_001243783.1 | NP_001230712.1 | 3 | 1522 | |||
| GenBank mRNA | AK025724.1 | - | 4 | 1435 | ||

BBIP1
Genomic Map