siRNA Details
| Gene Symbol: LINC00240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026775.2 | 1 | 177 | |||
| GenBank mRNA | BC031943.1 | - | 1 | 178 | ||
| BC044768.1 | - | 1 | 178 | |||
| BX647648.1 | - | 1 | 178 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00240| Gene Symbol: LINC00240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026775.2 | 1 | 177 | |||
| GenBank mRNA | BC031943.1 | - | 1 | 178 | ||
| BC044768.1 | - | 1 | 178 | |||
| BX647648.1 | - | 1 | 178 | |||