siRNA Details
| Gene Symbol: LINC00240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026775.2 | 3 | 425 | |||
| GenBank mRNA | BC031943.1 | - | 3 | 426 | ||
| BC044768.1 | - | 3 | 426 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00240| Gene Symbol: LINC00240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026775.2 | 3 | 425 | |||
| GenBank mRNA | BC031943.1 | - | 3 | 426 | ||
| BC044768.1 | - | 3 | 426 | |||