siRNA Details
| Gene Symbol: LOC641367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024523.1 | 1 | 536 | |||
| GenBank mRNA | BC024253.1 | - | 1 | 541 | ||
| BC047098.1 | - | 1 | 497 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC641367| Gene Symbol: LOC641367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024523.1 | 1 | 536 | |||
| GenBank mRNA | BC024253.1 | - | 1 | 541 | ||
| BC047098.1 | - | 1 | 497 | |||