siRNA Details
Gene Symbol: LOC641367 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024523.1 | 1 | 1394 | |||
GenBank mRNA | BC024253.1 | - | 1 | 1399 | ||
BC047098.1 | - | 1 | 1355 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY203313.1 | - | 1 | 68 | Chr. 19 - 21752100 - 21752544 [+] on Build GRCh38 |
