siRNA Details
Gene Symbol: TRAPPC2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002166.1 | 1, 2 | 193 | |||
GenBank mRNA | AF058918.1 | - | 1, 2 | 193 | ||
BU556832.1 | - | 1, 2 | 187 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TRAPPC2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002166.1 | 1, 2 | 193 | |||
GenBank mRNA | AF058918.1 | - | 1, 2 | 193 | ||
BU556832.1 | - | 1, 2 | 187 |