siRNA Details
Gene Symbol: FLJ34503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027060.1 | 2 | 453 | |||
GenBank mRNA | AK091822.1 | - | 2 | 702 | ||
BC093647.1 | - | 2 | 453 | |||
BC101497.1 | - | 2 | 453 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FLJ34503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027060.1 | 2 | 453 | |||
GenBank mRNA | AK091822.1 | - | 2 | 702 | ||
BC093647.1 | - | 2 | 453 | |||
BC101497.1 | - | 2 | 453 |