siRNA Details
| Gene Symbol: TCL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028288.1 | 4 | 1448 | |||
| GenBank mRNA | AB035337.1 | - | 4 | 1450 | ||
| BX161462.1 | - | 5 | 1113 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TCL6| Gene Symbol: TCL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028288.1 | 4 | 1448 | |||
| GenBank mRNA | AB035337.1 | - | 4 | 1450 | ||
| BX161462.1 | - | 5 | 1113 | |||