siRNA Details
Gene Symbol: TCL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028288.1 | 1 | 719 | |||
GenBank mRNA | AB035337.1 | - | 1 | 721 | ||
AB035338.1 | - | 1 | 721 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TCL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028288.1 | 1 | 719 | |||
GenBank mRNA | AB035337.1 | - | 1 | 721 | ||
AB035338.1 | - | 1 | 721 |