siRNA Details
Gene Symbol: LOC100133315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029192.1 | 5 | 585 | |||
GenBank mRNA | BX102226.1 | - | 4 | 395 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100133315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029192.1 | 5 | 585 | |||
GenBank mRNA | BX102226.1 | - | 4 | 395 |