siRNA Details
| Gene Symbol: SND1-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027330.1 | 1 | 1443 | |||
| GenBank mRNA | AF095687.1 | - | 1 | 1148 | ||
| AF191492.1 | - | 1 | 1443 | |||
| AK024176.1 | - | 1 | 1355 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SND1-IT1| Gene Symbol: SND1-IT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027330.1 | 1 | 1443 | |||
| GenBank mRNA | AF095687.1 | - | 1 | 1148 | ||
| AF191492.1 | - | 1 | 1443 | |||
| AK024176.1 | - | 1 | 1355 | |||