siRNA Details
| Gene Symbol: ABCC13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003087.1 | 4 | 592 | |||
| NR_003088.1 | 4 | 592 | ||||
| GenBank mRNA | AF418600.1 | - | 4 | 592 | ||
| AF518320.1 | - | 4 | 361 | |||
| AF518321.1 | - | 4 | 447 | |||
| AY063514.1 | - | 4 | 472 | |||
| AY063515.1 | - | 4 | 511 | |||
| AY344117.1 | - | 4 | 500 | |||
| BC144077.1 | - | 3 | 325 | |||

ABCC13
Genomic Map