siRNA Details
| Gene Symbol: RAI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033348.1 | 1 | 210 | |||
| NR_033349.1 | 1 | 210 | ||||
| XM_011545439.2 | XP_011543741.1 | 1 | 172 | |||
| XM_011545440.2 | XP_011543742.1 | 1 | 172 | |||
| XM_011545441.2 | XP_011543743.1 | 1 | 172 | |||
| GenBank mRNA | BC027937.1 | - | 1 | 90 | 5201821 | |
| DA377448.1 | - | 1 | 155 | |||
| DR005700.1 | - | 1 | 216 | |||

RAI2
Genomic Map