siRNA Details
Gene Symbol: LINC01348 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027454.2 | 3 | 745 | |||
GenBank mRNA | AK128489.1 | - | 2 | 2609 | ||
DA457760.1 | - | 3 | 562 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01348 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027454.2 | 3 | 745 | |||
GenBank mRNA | AK128489.1 | - | 2 | 2609 | ||
DA457760.1 | - | 3 | 562 |