siRNA Details
| Gene Symbol: FAM201A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027294.1 | 2 | 1323 | |||
| GenBank mRNA | BC036230.1 | - | 2 | 1322 | ||
| BC041970.1 | - | 2 | 1334 | |||
| BC067887.1 | - | 1 | 1240 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM201A| Gene Symbol: FAM201A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027294.1 | 2 | 1323 | |||
| GenBank mRNA | BC036230.1 | - | 2 | 1322 | ||
| BC041970.1 | - | 2 | 1334 | |||
| BC067887.1 | - | 1 | 1240 | |||