siRNA Details
| Gene Symbol: DISC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002227.2 | 1 | 1793 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AF222981.3 | - | 1 | 8222 | Chr. 1 - 231809945 - 231824936 [-] on Build GRCh38 | |

DISC2
Genomic Map