siRNA Details
| Gene Symbol: LINC00202-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026795.1 | 5 | 3576 | |||
| GenBank mRNA | AK097405.1 | - | 4 | 1743 | ||
| AK128027.1 | - | 4 | 5943 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX097650.1 | - | 3 | 490 | Chr. 10 - 26931211 - 26935497 [-] on Build GRCh38 | |

LINC00202-1
Genomic Map