siRNA Details
| Gene Symbol: LINC00202-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026795.1 | 1 | 1509 | |||
| GenBank mRNA | AK128027.1 | - | 1 | 2199 | ||
| Gene Symbol: ANKRD26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017015929.1 | XP_016871418.1 | 41 | 22697 | ||

LINC00202-1
Genomic Map