siRNA Details
| Gene Symbol: GAS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006721175.3 | XP_006721238.1 | 2, 3 | 162 | ||
| XM_011522991.1 | XP_011521293.1 | 3, 4 | 338 | |||
| XM_017023125.1 | XP_016878614.1 | 2, 3 | 162 | |||
| GenBank mRNA | AK303505.1 | - | 2, 3 | 165 | ||
| DC371501.1 | - | 2, 3 | 167 | |||

GAS8
Genomic Map