siRNA Details
| Gene Symbol: GBAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002188.3 | 5 | 455 | |||
| GenBank mRNA | BC030240.1 | - | 5 | 334 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GBAP1| Gene Symbol: GBAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002188.3 | 5 | 455 | |||
| GenBank mRNA | BC030240.1 | - | 5 | 334 | ||