siRNA Details
| Gene Symbol: SNHG20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027058.1 | 3 | 1115 | |||
| GenBank mRNA | AF130050.1 | - | 2 | 740 | ||
| BX648413.1 | - | 3 | 1075 | |||
| FN357117.1 | - | 3 | 1009 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNHG20| Gene Symbol: SNHG20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027058.1 | 3 | 1115 | |||
| GenBank mRNA | AF130050.1 | - | 2 | 740 | ||
| BX648413.1 | - | 3 | 1075 | |||
| FN357117.1 | - | 3 | 1009 | |||