siRNA Details
| Gene Symbol: LINC00221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027457.2 | 3 | 721 | |||
| GenBank mRNA | BX161407.1 | - | 2 | 524 | ||
| BX248748.1 | - | 3 | 693 | |||
| BX356743.2 | - | 3 | 647 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00221| Gene Symbol: LINC00221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027457.2 | 3 | 721 | |||
| GenBank mRNA | BX161407.1 | - | 2 | 524 | ||
| BX248748.1 | - | 3 | 693 | |||
| BX356743.2 | - | 3 | 647 | |||