siRNA Details
| Gene Symbol: LINC00221 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027457.2 | 3 | 900 | |||
| GenBank mRNA | BX161407.1 | - | 2 | 703 | ||
| BX248748.1 | - | 3 | 872 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BX333112.2 | - | 3 | 771 | Chr. 14 - 106482569 - 106494867 [+] on Build GRCh38 | |

LINC00221
Genomic Map