siRNA Details
| Gene Symbol: LINC00475 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027341.1 | 2 | 214 | |||
| GenBank mRNA | AK023662.1 | - | 2 | 214 | ||
| BC128384.1 | - | 2 | 99 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00475| Gene Symbol: LINC00475 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027341.1 | 2 | 214 | |||
| GenBank mRNA | AK023662.1 | - | 2 | 214 | ||
| BC128384.1 | - | 2 | 99 | |||