siRNA Details
| Gene Symbol: ZNF295-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027273.2 | 1, 2 | 170 | |||
| NR_119384.1 | 1, 2 | 399 | ||||
| GenBank mRNA | BC029588.1 | - | 1, 2 | 170 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF295-AS1| Gene Symbol: ZNF295-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_027273.2 | 1, 2 | 170 | |||
| NR_119384.1 | 1, 2 | 399 | ||||
| GenBank mRNA | BC029588.1 | - | 1, 2 | 170 | ||