siRNA Details
Gene Symbol: LINC00202-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026794.1 | 1 | 3883 | |||
GenBank mRNA | CR749234.1 | - | 1 | 3823 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00202-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026794.1 | 1 | 3883 | |||
GenBank mRNA | CR749234.1 | - | 1 | 3823 |