siRNA Details
| Gene Symbol: PPBPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026769.1 | 1 | 576 | |||
| GenBank mRNA | BC132938.1 | - | 1 | 499 | ||
| BC132940.1 | - | 1 | 498 | |||
| L10403.1 | - | 1 | 576 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPBPP2| Gene Symbol: PPBPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026769.1 | 1 | 576 | |||
| GenBank mRNA | BC132938.1 | - | 1 | 499 | ||
| BC132940.1 | - | 1 | 498 | |||
| L10403.1 | - | 1 | 576 | |||