siRNA Details
| Gene Symbol: LINC00705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015425.1 | 1 | 86 | |||
| GenBank mRNA | BC040895.1 | - | 1 | 815 | ||
| BC101460.1 | - | 1 | 86 | |||
| BC101989.1 | - | 1 | 86 | |||
| BC104742.1 | - | 1 | 86 | |||
| BC129979.1 | - | 1 | 86 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000417883.2 | - | 1 | 815 | Chr. 10 - 4655427 - 4662419 [+] on Build GRCh38 | |

LINC00705
Genomic Map