siRNA Details
| Gene Symbol: LINC00705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015425.1 | 3 | 955 | |||
| GenBank mRNA | BC040895.1 | - | 4 | 1684 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00705| Gene Symbol: LINC00705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015425.1 | 3 | 955 | |||
| GenBank mRNA | BC040895.1 | - | 4 | 1684 | ||