siRNA Details
| Gene Symbol: LINC00705 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015425.1 | 2 | 210 | |||
| GenBank mRNA | BC040895.1 | - | 2 | 939 | ||
| BC101460.1 | - | 2 | 210 | |||
| BC101989.1 | - | 2 | 210 | |||
| BC104742.1 | - | 2 | 210 | |||
| BC129979.1 | - | 2 | 210 | |||

LINC00705
Genomic Map