siRNA Details
| Gene Symbol: LINC00961 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024283.1 | 2 | 923 | |||
| GenBank mRNA | AK056723.1 | - | 2 | 923 | ||
| BC034765.1 | - | 1 | 922 | |||
| BC113682.1 | - | 1 | 784 | |||
| BC113684.1 | - | 1 | 784 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00961| Gene Symbol: LINC00961 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024283.1 | 2 | 923 | |||
| GenBank mRNA | AK056723.1 | - | 2 | 923 | ||
| BC034765.1 | - | 1 | 922 | |||
| BC113682.1 | - | 1 | 784 | |||
| BC113684.1 | - | 1 | 784 | |||