siRNA Details
Gene Symbol: LINC00895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024381.1 | 1 | 636 | |||
GenBank mRNA | AK057431.1 | - | 1 | 636 | ||
BC104250.1 | - | 1 | 578 | |||
BC104251.1 | - | 1 | 575 | |||
BC113002.1 | - | 1 | 578 | |||
BC113003.1 | - | 1 | 578 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG721941.1 | - | 1 | 639 | Chr. 22 - 19564961 - 19566843 [-] on Build GRCh38 |
