siRNA Details
| Gene Symbol: LINC00895 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024381.1 | 1 | 663 | |||
| GenBank mRNA | AK057431.1 | - | 1 | 663 | ||
| BC104250.1 | - | 1 | 605 | |||
| BC104251.1 | - | 1 | 602 | |||
| BC113002.1 | - | 1 | 605 | |||
| BC113003.1 | - | 1 | 605 | |||

LINC00895
Genomic Map