siRNA Details
| Gene Symbol: ATP8B5P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_003581.2 | 10 | 842 | |||
| NR_003582.1 | 10 | 846 | ||||
| NR_110320.1 | 10 | 842 | ||||
| GenBank mRNA | AK310278.1 | - | 10 | 818 | ||
| BC031276.1 | - | 10 | 829 | |||
| BC035229.1 | - | 9 | 2405 | |||
| BC130448.1 | - | 5 | 606 | |||
| BC130450.1 | - | 5 | 606 | |||

ATP8B5P
Genomic Map