siRNA Details
| Gene Symbol: LINC00896 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026919.1 | 1 | 337 | |||
| GenBank mRNA | AK057137.1 | - | 1 | 272 | ||
| CR456358.1 | - | 1 | 92 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00896| Gene Symbol: LINC00896 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026919.1 | 1 | 337 | |||
| GenBank mRNA | AK057137.1 | - | 1 | 272 | ||
| CR456358.1 | - | 1 | 92 | |||