siRNA Details
| Gene Symbol: TTTY6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_001527.2 | 2 | 129 | |||
| Gene Symbol: TTTY6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_002175.3 | 2 | 107 | |||

TTTY6
Genomic Map