siRNA Details
| Gene Symbol: KRTAP5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_021489.1 | 2 | 2097 | |||
| GenBank mRNA | AK055141.1 | - | 2 | 1788 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP5-AS1| Gene Symbol: KRTAP5-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_021489.1 | 2 | 2097 | |||
| GenBank mRNA | AK055141.1 | - | 2 | 1788 | ||