siRNA Details
| Gene Symbol: WDFY3-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015359.1 | 4 | 831 | |||
| GenBank mRNA | AY250185.1 | - | 7 | 1354 | ||
| BC019264.1 | - | 7 | 907 | |||
| BC033127.1 | - | 4 | 812 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

WDFY3-AS2| Gene Symbol: WDFY3-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015359.1 | 4 | 831 | |||
| GenBank mRNA | AY250185.1 | - | 7 | 1354 | ||
| BC019264.1 | - | 7 | 907 | |||
| BC033127.1 | - | 4 | 812 | |||