siRNA Details
| Gene Symbol: MORC2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026920.1 | 1 | 95 | |||
| GenBank mRNA | BC042980.1 | - | 1 | 57 | ||
| DB038414.1 | - | 1 | 95 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MORC2-AS1| Gene Symbol: MORC2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026920.1 | 1 | 95 | |||
| GenBank mRNA | BC042980.1 | - | 1 | 57 | ||
| DB038414.1 | - | 1 | 95 | |||