siRNA Details
| Gene Symbol: ZNF577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024181.1 | 8 | 922 | |||
| GenBank mRNA | AK056853.1 | - | 3 | 1785 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | DB527989.1 | - | 5 | 464 | Chr. 19 - 51855804 - 51880339 [-] on Build GRCh38 | |

ZNF577
Genomic Map