siRNA Details
| Gene Symbol: ZNF577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024181.1 | 8 | 803 | |||
| GenBank mRNA | AK056853.1 | - | 3 | 1666 | ||
| AK093036.1 | - | 7 | 1738 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF577| Gene Symbol: ZNF577 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024181.1 | 8 | 803 | |||
| GenBank mRNA | AK056853.1 | - | 3 | 1666 | ||
| AK093036.1 | - | 7 | 1738 | |||