siRNA Details
| Gene Symbol: TYRO3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028510.1 | 1 | 719 | |||
| GenBank mRNA | X72887.1 | - | 1 | 719 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TYRO3P| Gene Symbol: TYRO3P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_028510.1 | 1 | 719 | |||
| GenBank mRNA | X72887.1 | - | 1 | 719 | ||